missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Flat Sealing Tapes

Shapeable PTFE flat sealing tapes.

Brand:  Bohlender™ H959-34

Product Code. 17107424

  • 354.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Deformable, virgin PTFE flat tape with expanded fibre structure
  • Tasteless
  • Odorless up to 270°C
  • Self-adhesive
  • Physiologically safe
  • Not ageing
  • Good sealing even on uneven surfaces
  • Almost universal chemical resistance
  • Quick and easy assembly
Specifications

Specifications

0.2 in.
7 mm (0.28 in.)
PTFE
5 mm
22 mm
0.87 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers