Learn More
Bohlender™ BOLA™ Measuring Cylinders
Translucent measuring cylinders with hexagonal base to prevent rolling.
Brand: Bohlender™ A164-30
This item is not returnable.
View return policy
Description
- Translucent and non-porous
- Design based on ISO 4788
- With graduation (uncalibrated), spout and reinforcing ring
- Universal chemical resistance
- Product is only exposed to PTFE
- Hexagonal base prevents the cylinder from rolling
- Permanent, recessed graduation
Specifications
500 mL | |
5 mL | |
Cylinder | |
2.1 in. | |
308 mm |
PTFE | |
17 oz. | |
53 mm | |
12.1 in. |