missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Miniature 3-Way Stopcocks, PTFE, FEP

Connect a wide range of tubing sizes with these stopcocks. BOLA Minature Three-way stopcocks feature universal chemical resistance.

Brand:  Bohlender™ F731-08

Product Code. 10297401

  • 174.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 3-way stopcock with “L”-shaped or “T”-shaped bore and three connections
  • Available either with three female threads UNF 1/4 in. 28 G or with two female threads UNF 1/4 in. 28 G and one male thread UNF1/4 in. 28 G
  • Total height including stopcock plug made of FEP: 20mm
Specifications

Specifications

FDA
Autoclavable
-200°C
3 female thread connections
White/Red
250°C
Miniature 3-Way Stopcock
32 mm
1.6 x 3.2mm (I.D. x O.D.)
T-shape
8 bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.