Learn More
Bohlender™ BOLA™ PTFE/PPS HT Replacement Inner Parts
Consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3mm (not exposed to the medium).
26.00 € - 199.00 €
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Material PPS, PTFE
Min. Operating Temperature (Metric) -200°C
Color White
View More Specs
Products 14
Product Code Brand Tubing Size Thread Size Price Quantity & Availability  
Product Code Brand Tubing Size Thread Size Price Quantity & Availability  
10409391
View Documents
Bohlender™
D639-22
2mm O.D. GL 18
26.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10616683
View Documents
Bohlender™
D640-42
(1/8 in.) 3.2mm O.D. GL 25
46.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10043371
View Documents
Bohlender™
D640-18
(1/16 in.) 1.6mm O.D. GL 25
32.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10267451
View Documents
Bohlender™
D642-74
(1 in.) 25.4mm O.D. GL 45
118.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10651522
View Documents
Bohlender™
D640-46
4mm O.D. GL 25
37.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10134921
View Documents
Bohlender™
D641-38
6mm O.D. GL 32
107.00 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
10491872
View Documents
Bohlender™
D640-22
2mm O.D. GL 25
31.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10368941
View Documents
Bohlender™
D641-60
12mm O.D. GL 32
107.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10124971
View Documents
Bohlender™
D641-56
10mm O.D. GL 32
100.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10504212
View Documents
Bohlender™
D641-42
(1/4 in.) 6.35mm O.D. GL 32
199.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10449191
View Documents
Bohlender™
D641-78
16mm O.D. GL 32
99.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10575742
View Documents
Bohlender™
D642-44
12mm O.D. GL 45
142.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10184631
View Documents
Bohlender™
D642-76
26mm O.D. GL 45
97.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10144921
View Documents
Bohlender™
D641-46
8mm O.D. GL 32
102.00 €
Each
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
Description
Description
  • Medium is only exposed to PTFE
  • Good chemical resistance
  • Withstand pressure up to 10 bar
  • Vacuum suitable
Specifications
Specifications
FDA
PPS, PTFE
White
250°C
Autoclavable
-200°C
10 bar