Learn More
Bohlender™ Screw Caps with Flexible Distributors Product Code.: 10267594
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Bohlender™ Screw Caps with Flexible Distributors
Product Code. 10267594
Tubing I.D. 7-11mm
Click to view available options
For Use With (Equipment):
Tubing I.D. 7-11mm
Product Code. 10267594
Brand: Bohlender™ D80048
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Add to basket
Use these caps for drawing or inserting liquids. Bohlender™ Screw Caps with Flexible Distributors are made of green polypropylene for bottle thread GL 45 and distributor body with hose connectors made of polypropylene.
  • Bent hose connectors on the upper side
  • Straight hose connectors on the bottom side
  • Restricted chemical resistance
  • For working temperatures up to max. +110°C
Specifications
For Use With (Application) Drawing or inserting liquids, for elastic tubing (e.g. Viton™, Tygon™, silicone)
Temperature Range (Metric) -20 to 110 deg.C
Closure Color Green
Product Type Screw Cap with Flexible Distributors
For Use With (Equipment) Tubing I.D. 7-11mm
Product Content Correction
Product Title
Bohlender™ Screw Caps with Flexible Distributors >