Learn More
Bohlender™ BOLA™ Spare O-rings for Stopcocks
Spare o-rings for stopcocks.
Brand:  Bohlender™ J1198-01
Product Code. 17107474
  • 38.43 € / Pack of 20
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • O-rings made of NBR
  • For BOLA™ Eco Stopcock Plugs
  • Taper: 1:10
  • Thread M: 7 mm
  • Suitable for size: NS 10
Specifications
Specifications
Spare O-ring
NBR
For BOLA™ Eco Stopcock Plugs