Learn More
Bohlender™ BOLA™ Spare V-rings, Two-part Set
Set of two spare v-rings.
Brand:  Bohlender™ D502-06
 View more versions of this product
Product Code. 17197194
  • 13.00 € / Pack of 5
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of PTFE
  • Two-part set with one tapered ring and one v-ring
Specifications
Specifications
PTFE
10 mm
M 18 x 2
One Tapered Ring, One V-ring