Learn More
Bohlender™ BOLA Standard Construction Kit
For making flanges at the ends of plastic tubing (ex. PTFE, PFA or FEP)
Brand:  Bohlender™ F700-04
Product Code. 10003551
  • 622.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Consisting of:
    • 1x BOLA Thermoelectric Flanging Tool with exchangeable flanging tip for tubing I.D. 1.6mm and tubing holder for tubing O.D. 1.6mm (1/16 in.) and 3.2 mm (1/8 in.)
    • 1 x BOLA Tube End Fitting Set
    • 5 x BOLA Plugs
    • 10 x BOLA Miniature Couplings (straight)
    • 2 x Miniature Couplings (T)
    • 1 x BOLA Miniature Coupling (cross)
    • 10 metres of PTFE tubing (I.D. 1.6mm, O.D. 3.2mm)
  • Specifications
    Specifications
    BOLA Standard Construction Kit
    Not Autoclavable
    For making flanges at the ends of plastic tubing
    BOLA Standard Construction Kit
    Plastic tubing