missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Empty Mini Gel Cassette Combs, 12-well

Used with Bolt™ Empty Minigel Cassettes (Mfr. No. NW2010) for use in gel electrophoresis. Bolt™ Minigel Cassette Combs give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank.

Brand:  Invitrogen™ NW3012

21.49 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15314614

  • 24.99 € / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price


Includes: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy

Description

Description

Bolt Empty Gel Cassette Combs are for use with Bolt Empty Gel Cassettes, which give you the option of pouring your own special gel type while still enjoying the advantages of the Mini Gel Tank and XCell SureLock Mini-Cell. All cassettes are sealed on three sides (so no gel leakage) and feature the unique wedge-shaped well design. The cassettes and combs are available in the standard 1-mm thickness. The combs are available for 10 or 12 wells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mini Gel Tank and XCell SureLock Mini-Cell
Bolt
20 combs
Mini (8 x 8 cm)
Empty Mini Gel Cassette Comb
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.