Learn More
Macherey-Nagel™ BondEX™ Starter Kit
For decontamination and removal of ethidium boride or SYBR™ Green
Brand:  Macherey-Nagel™ 740701
Product Code. 10289591
  • 221.00 € / Each
Explore more special offers
Description
Description
Decontamination and removal of ethidium bromide or SYBR™ Green waste is often time consuming and expensive. bondEX EtBr cartridges save time and money as the prefiltered ethidium bromide or SYBR™ Green containing solution is simply poured onto the cartridge resulting in a flow-through free of ethidium bromide or SYBR™ Green (clarification filters included in the kit). For larger volumes a 1L funnel can be connected to the top of the cartridge. Up to 50mg of ethidium bromide can be adsorbed onto the bondEX EtBr cartridge. A removable indicator cartridge signals saturation of the bondEX cartridge by red color or UV fluorescence. The bondEX EtBr cartridges are recommended for the extraction of ethidium bromide from diluted EtBr solutions like TAE or TBE gel buffer as well as for more concentrated solutions. Similar to ethidium bromide, SYBR™ Green is a highly sensitive fluorescent stain for detecting nucleic acids in agarose and polyacrylamide gels. According to the manufacturer SYBR™ Green is less mutagenic than ethidium bromide. The problem arises how to store and decontaminate SYBR™ Green waste and SYBR™ Green containing solutions. Solutions can be passed through a bondEX EtBr cartridge as described for ethidium bromide waste. The binding capacity for SYBR™ Green is comparable to EtBr. Kit components: bondEX starter kit – 2 cartridges (50mg EtBr capacity each), 6 indicator cartridges, 1L funnel and adapter, 1 adapter for connection to container, 10 folded filters and 1 plastic funnel. bondEX 50 – each set consists of 1 cartridge (50mg EtBr capacity) with 2 end caps, hazard label, supporting ring, 3 indicator cartridges, 2 folded filters.
  • High capacity - up to 50mg EtBr or SYBR™ Green can be bound to the resin
  • Safe handling - no contamination, cartridges can be closed
  • Indicator cartridge with red color or UV fluorescence signalizes saturation
  • No interference by other dyes like methylene blue, because indicator cartridges can be replaced
  • Time saving procedure - the cartridges run by gravity, no additional tools, such as syringes, pumps etc. are necessary
  • Convenience for large volumes - a 1L funnel (included in starter kit) or a bigger reservoir can easily be connected
    Product at a glance
    Technology: HIC (hydrophobic interaction chromatography)Format: Gravity-flow columnsSample material: <50L filtered solutions containing up to 50mg EtBr or SYBR™ GreenFiltration of solutions: bondEX Folded Filters XL (provided)Binding capactity: 50mg EtBr or SYBR™ Green For decontamination of solutions containing fluorescent staining agents.
Specifications
Specifications
BondEX Starter Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products