missing translation for 'onlineSavingsMsg'
Learn More

S Murray and Co™ Bone Cutter

Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Bone Cutters are fully guaranteed in normal use and subject to stringent quality inspection procedures.

Brand:  S Murray and Co™ D632/01

Product Code. 12734416

  • 87.50 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Bone Cutters are fully guaranteed in normal use and subject to stringent quality inspection procedures.

  • Stainless Steel
  • Length: 20mm
Specifications

Specifications

20 mm
1 Ea.
Bone Cutter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S Murray and Co™ Bone Cutter > Length: 20mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.