missing translation for 'onlineSavingsMsg'
Learn More

Borane-d{3}, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

227.13 € - 561.00 €

Chemical Identifiers

CAS 13763-62-7
Molecular Formula BD3
Molecular Weight (g/mol) 16.85
MDL Number MFCD00064892
InChI Key UORVGPXVDQYIDP-FIBGUPNXSA-N
Synonym borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles
PubChem CID 11062299
IUPAC Name trideuterioborane
SMILES [2H]B([2H])[2H]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15421977
SDS View Documents
Thermo Scientific Alfa Aesar
044188.AB
10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15431977
SDS View Documents
Thermo Scientific Alfa Aesar
044188.AD
50 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H302-H314-H335-H336-H351

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Borane-d{3}, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.