Learn More
Borane-d{3}, 1M in tetrahydrofuran, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 13763-62-7 |
---|---|
Molecular Formula | BD3 |
Molecular Weight (g/mol) | 16.85 |
MDL Number | MFCD00064892 |
InChI Key | UORVGPXVDQYIDP-FIBGUPNXSA-N |
Synonym | borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles |
PubChem CID | 11062299 |
SMILES | [2H]B([2H])[2H] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13763-62-7 | |
16.85 | |
UORVGPXVDQYIDP-FIBGUPNXSA-N | |
11062299 |
BD3 | |
MFCD00064892 | |
borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles | |
[2H]B([2H])[2H] |
Specifications
13763-62-7 | |
Liquid | |
MFCD00064892 | |
-21°C (-6°F) | |
UORVGPXVDQYIDP-FIBGUPNXSA-N | |
trideuterioborane | |
11062299 | |
1M in tetrahydrofuran | |
Air & Moisture Sensitive | |
Borane-d_3 |
Colorless | |
BD3 | |
UN3148 | |
borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles | |
[2H]B([2H])[2H] | |
16.85 | |
16.84 as BD3 | |
Ether-like | |
0.876g/mL |
Safety and Handling
GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335-H336-H351
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.