missing translation for 'onlineSavingsMsg'
Learn More

Borane dimethyl sulfide complex, 94%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10275750
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-87-0, 75-18-3
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name boron;methylsulfanylmethane
SMILES B.CSC

Specifications

Melting Point -40°C to 37°C
Color Amber to Colorless
Density 0.8010g/mL
Flash Point 3°C
Assay Percent Range 94%
Packaging AcroSeal™ Glass bottle
Linear Formula (CH3)2S·BH3
Quantity 100 mL
Fieser 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Merck Index 15, 1336
Specific Gravity 0.801
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ethyl ether,tetrahydrofurane,hexane,,toluene,methylene chloride and other aprotic,solvents
Formula Weight 75.95
Percent Purity 94%
Physical Form Solution
Chemical Name or Material Borane dimethyl sulfide complex, 94%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Borane-methyl sulfide complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H301-Toxic if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'