Learn More
Borane-methyl sulfide complex, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 185071000
73.13 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane-methyl sulfide complex | |
2M Solution in Tetrahydrofuran | |
C2H9BS | |
MFCD00013189 | |
04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50 | |
15, 1336 | |
Solubility in water: reacts. Other solubilities: soluble in ethyl ether, tetrahydrofurane, hexane,, toluene, methylene chloride and other aprotic, solvents | |
B.CSC | |
75.96 | |
75.95 | |
0.85 | |
−17°C |
Colorless to Yellow | |
13292-87-0, 109-99-9, 75-18-3 | |
(CH3)2S·BH3 | |
1.80 to 2.20M | |
0.8500g/mL | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
(methylsulfanyl)methane borane | |
9833925 | |
AcroSeal™ Glass bottle | |
100 mL | |
Solution |
Safety and Handling
- Borane-methyl sulfide complex, 2M solution in tetrahydrofuran
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only