missing translation for 'onlineSavingsMsg'
Learn More

Borane dimethyl sulfide complex, 2M solution in THF, AcroSeal™

Product Code. p-71779 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10022420

Brand: Thermo Scientific Acros 185071000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Borane-methyl sulfide complex
Color Colorless to Yellow
Name Note 2M Solution in Tetrahydrofuran
CAS 13292-87-0, 109-99-9, 75-18-3
Molecular Formula C2H9BS
Linear Formula (CH3)2S·BH3
MDL Number MFCD00013189
Concentration 1.80 to 2.20M
Fieser 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Density 0.8500g/mL
Merck Index 15, 1336
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ethyl ether, tetrahydrofurane, hexane,, toluene, methylene chloride and other aprotic, solvents
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES B.CSC
IUPAC Name (methylsulfanyl)methane borane
Molecular Weight (g/mol) 75.96
PubChem CID 9833925
Formula Weight 75.95
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.85
Quantity 100 mL
Flash Point −17°C
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Borane-methyl sulfide complex, 2M solution in tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.