missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 8.0
64.82 € - 96.48 €
Chemical Identifiers
CAS | 1330-43-4 |
---|---|
Molecular Formula | B4Na2O7 |
Molecular Weight (g/mol) | 201.21 |
MDL Number | MFCD00081185 |
InChI Key | NXSVMNYRVBQCMO-UHFFFAOYSA-N |
IUPAC Name | disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
SMILES | [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425929
|
Thermo Scientific Chemicals
J63742.AK |
250 mL |
64.82 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15435929
|
Thermo Scientific Chemicals
J63742.AP |
500 mL |
96.48 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1330-43-4 | |
201.21 | |
NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
B4Na2O7 | |
MFCD00081185 | |
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
Specifications
1330-43-4 | |
8.0 | |
B4Na2O7 | |
NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) | |
Borate buffer soln. |
0.5 M | |
Liquid | |
MFCD00081185 | |
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 | |
201.21 |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H360FD
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only