missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid 4% w/v with Indicator Kjeldahl Reagent, Reagecon
BIOPHARMA

Boric Acid 4% w/v with Indicator Kjeldahl Reagent

Brand:  Reagecon KJR015

Product Code. 11503192

  • 101.00 € / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Boric Acid 4% w/v with Indicator Kjeldahl Reagent
Specifications

Specifications

5 L
HDPE Container
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberKJR0123G1 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog NumberKJR015
Lot NumberKJR0122D1 Certificate TypeCertificate of Analysis Date23/02/2023 Catalog NumberKJR015
Lot NumberKJR0120G1 Certificate TypeCertificate of Analysis Date28/10/2021 Catalog NumberKJR015
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boric Acid 4% w/v with Indicator Kjeldahl Reagent, Reagecon > 5 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.