missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 98%, Thermo Scientific Chemicals

Product Code. 11447967
Click to view available options
Quantity:
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity

Solubility
Soluble in glycerol, ether, alcohol, methanol, liquid ammonia, acetone (slightly), water (6.35 g/100 cc) at 30°C, water (27.6 g/100 cc) at 100°C, water (50 g/L) at 21°C, and water (1 g / 18 ml cold water).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Melting Point ∼185°C (decomposition)
Density 1.435
Odor Odorless
Flash Point None
Quantity 5000 g
Linear Formula H3BO3
Beilstein 1697939
Merck Index 14,1336
Solubility Information Soluble in glycerol,ether,alcohol,methanol,liquid ammonia,acetone (slightly),water (6.35g/100 cc) at 30°C,water (27.6g/100 cc) at 100°C,water (50g/L) at 21°C,and water (1 g / 18 ml cold water).
Formula Weight 61.83
Percent Purity 98%
Chemical Name or Material Boric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.