missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Certified AR for Analysis, Fisher Chemical™
65.13 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Description
H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 3.8 to 4.8 |
Color | White |
Melting Point | 169°C |
Packaging | Plastic powder jar |
Vapor Pressure | 2.7mbar at 20°C |
Quantity | 1 kg |
Physical Form | Powder Solid |
Formula Weight | 61.83g/mol |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : boric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'