missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Powder 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
Color | White |
Melting Point | 169.0°C |
Odor | Odorless |
Packaging | Plastic bottle |
Assay Percent Range | 99+% |
Quantity | 2.5 kg |
Linear Formula | H3BO3 |
Physical Form | Coarse Crystals |
Fieser | 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70 |
Merck Index | 15, 1339 |
Show More |
Safety and Handling
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Boric acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction