missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Vapor Pressure | 2.6mmHg (20°C) |
Quantity | 2.5 kg |
Linear Formula | H3BO3 |
UN Number | NONH for all modes of transport |
Solubility Information | In accordance with ethanol |
Formula Weight | 61.83g/mol |
Percent Purity | 99.5 to 100.5% |
Grade | puriss. |
Chemical Name or Material | Boric Acid, Meets analytical specification of Ph. Eur.BP, NF |
Safety and Handling
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : boric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction