Learn More
BRAND™ Boro 3.3 Conical Ground Joint Stopper
Solid or semi-hollow, with octagonal grip
Brand:  BRAND™ 144530
10.40 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10597451
  • 12.05 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Boro 3.3 glass construction
  • DIN 12252
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Solid
    NS 24/29
    Borosilicate Glass
    Octagonal Grip