Learn More
Boron carbide, Thermo Scientific Chemicals
49.70 € - 178.00 €
Chemical Identifiers
CAS | 12069-32-8 |
---|---|
Molecular Formula | CB4 |
Molecular Weight (g/mol) | 55.25 |
MDL Number | MFCD00011520 |
InChI Key | INAHAJYZKVIDIZ-UHFFFAOYSA-N |
Synonym | boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07 |
PubChem CID | 123279 |
IUPAC Name | 2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane |
SMILES | B12B3B4B1C234 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12069-32-8 | |
55.25 | |
INAHAJYZKVIDIZ-UHFFFAOYSA-N | |
123279 | |
B12B3B4B1C234 |
CB4 | |
MFCD00011520 | |
boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07 | |
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane |
Specifications
2350°C | |
Powder | |
MFCD00011520 | |
boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07 | |
B12B3B4B1C234 | |
55.25 | |
55.26 | |
2.52 g/mL |
12069-32-8 | |
CB4 | |
14,1344 | |
INAHAJYZKVIDIZ-UHFFFAOYSA-N | |
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane | |
123279 | |
Odorless | |
Boron carbide |
Safety and Handling
GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H332-H335
missing translation for 'einecsNumber' : 235-111-5
missing translation for 'rtecsNumber' : ED7420000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.