missing translation for 'onlineSavingsMsg'
Learn More
Boron nitride, 98%, Thermo Scientific Chemicals Product Code.: 15580544

Boron nitride, 98%, Thermo Scientific Chemicals

Product Code. 15580544
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 15580544

Brand: Thermo Scientific Acros 441800500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Melting Point 2700.0°C to 3000.0°C
Quantity 50 g
Assay Percent Range 97.5% min
Linear Formula BN
Merck Index 15,1347
Solubility Information Solubility in water: slightly soluble (20°C). Other solubilities: soluble in water (95°)
IUPAC Name boranylidyneamine
Formula Weight 24.82
Percent Purity 98%
Chemical Name or Material Boron nitride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461451 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number441800500,441802500
Lot NumberA0453544 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number441800500,441802500
Lot NumberA0403031 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number441800500,441802500
Lot NumberA0392534 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number441800500,441802500
Lot NumberA0407622 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number441800500,441802500
Product Identifier
  • Boron nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron nitride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.