missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals

129.65 € - 435.17 €

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386088
SDS View Documents
Thermo Scientific Alfa Aesar
011078.18
50 g
129.65 €
50g
Please to purchase this item. Need a web account? Register with us today!
11396088
SDS View Documents
Thermo Scientific Alfa Aesar
011078.30
250 g
435.17 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Boron nitride used in the manufacture of high-temperature equipment parts due to its excellent thermal and chemical stability. It is used as a lubricant and an additive to cosmetics, paints, dental cements and pencil leads. It is also used to provide self lubricating properties to ceramics, alloys, resins, plastics and rubbers. In addition to this, it has many industrial applications such as xerographic process, laser printers, oxygen sensors and proton conductors. Cubic boron nitride is used as an abrasive material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10043-11-5
24.82
LNLSXDSWJBUPHM-UHFFFAOYSA-N
66227
B=N
BN
MFCD00011317
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
CHEBI:50883

Specifications

10043-11-5
99.5% (metals basis)
MFCD00011317
Insoluble in water.
B=N
24.82
CHEBI:50883
Boron nitride
Powder
BN
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
LNLSXDSWJBUPHM-UHFFFAOYSA-N
azanylidyneborane
66227
24.82
Videos
Safety and Handling

Safety and Handling

P261-P271-P304+P340-P312-P501c

H335

missing translation for 'einecsNumber' : 233-136-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.