missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 99.5% (metals basis), Thermo Scientific™

Product Code. 15422047
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranylidyneamine
SMILES B=N

Specifications

Physical Form Hot Pressed Rod
Quantity 5 Pc.
Solubility Information Insoluble in water.
Formula Weight 24.82
Percent Purity 99.5%
Packaging Bag
Chemical Name or Material Boron nitride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.