missing translation for 'onlineSavingsMsg'
Learn More
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 15452377

Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 15452377
1 Pc.
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy

Product Code. 15452377

Brand: Thermo Scientific Alfa Aesar 045575.KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.

Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranylidyneamine
SMILES B=N

Specifications

Physical Form Hot Pressed Rod
Quantity 1 Pc.
Solubility Information Insoluble in water. Boron nitride is insoluble in the acids. Soluble in alkaline molten salts and nitrides,
Formula Weight 24.82
Percent Purity 99.5%
Packaging Bag
Chemical Name or Material Boron nitride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL01T050 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045575.KG
Lot NumberJ08Z044 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045575.KG
Lot NumberN19B022 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045575.KG
Lot NumberP21C015 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045575.KG

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.