missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride paste, Thermo Scientific Chemicals

Product Code. 11370408
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Has Resistance Against Molten Metals and Salts, High thermal Conductivity and Dielectric Strength

Resistance against molten metals and salts, high thermal conductivity and dielectric strength

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Color White
Physical Form Paste
Quantity 2 kg
IUPAC Name boranimine
Odor Odorless
Appearance White paste
Density 1.4
Chemical Name or Material Boron nitride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'