missing translation for 'onlineSavingsMsg'
Learn More
Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals Product Code.: 11311649

Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals

Product Code. 11311649
2 kg
Click to view available options
Quantity:
500 g
2 kg
10 kg
This item is not returnable. View return policy

Product Code. 11311649

Brand: Thermo Scientific Alfa Aesar 012290.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from Talc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500gShow MoreShow Less
PubChem CID 518682
ChEBI CHEBI:30163
SMILES O=BOB=O

Specifications

Boiling Point 1860°C
Melting Point ∽450°C
Quantity 2 kg
Assay Percent Range 97.5% min., typically 98.5%
Solubility Information Slightly soluble in water and methanol.
IUPAC Name oxo(oxoboranyloxy)borane
Formula Weight 69.62
Sensitivity Hygroscopic
Density 2.46
Chemical Name or Material Boron oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW13K026 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number012290.A3,012290.36,012290.A9
Lot NumberW12I022 Certificate TypeCertificate of Analysis Date16/09/2022 Catalog Number012290.A3,012290.36,012290.A9
Lot NumberY16G050 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog Number012290.A3,012290.36,012290.A9
Lot NumberT10G002 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012290.A3,012290.36,012290.A9
Lot NumberU25C014 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012290.A3,012290.36,012290.A9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.