Learn More
Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1860°C |
Melting Point | ∽450°C |
Color | White |
Physical Form | Powder |
Quantity | 2 g |
Assay Percent Range | 99.999% |
Solubility Information | Soluble in water. Slightly soluble in methanol. |
Formula Weight | 69.62 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only