missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals

For Use in the Electronics Industry As a Liquid Encapsulent For Czochralski-Type Growth of III-V Semiconductor Single Crystals (E.G., Gaas, Gap

105.53 € - 831.14 €

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
SMILES O=BOB=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396178
SDS View Documents
Thermo Scientific Alfa Aesar
011159.QE
32 g
105.53 €
Each
Please to purchase this item. Need a web account? Register with us today!
11386178
SDS View Documents
Thermo Scientific Alfa Aesar
011159.QH
10 x 32 g
831.14 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

In the electronics industry as a liquid encapsulant for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-86-2
69.62
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
O=BOB=O
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163

Specifications

1860°C
1303-86-2
Cylinder
B2O3
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo[(oxoboranyl)oxy]borane
518682
69.62
Hygroscopic
2.46±0.01 g/cm3 at 20°C
∽450°C
White
99.999% (metals basis)
MFCD00011315
Soluble in water. Slightly soluble in methanol.
O=BOB=O
69.62
CHEBI:30163
Odorless
White cylinder
Boron oxide, (H2O 200ppm)
Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P201-P202-P281-P308+P313-P501c

H360FD

missing translation for 'einecsNumber' : 215-125-8

missing translation for 'rtecsNumber' : ED7900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.