Learn More
Boron phosphate, Thermo Scientific Chemicals
143.72 € - 581.90 €
Chemical Identifiers
CAS | 13308-51-5 |
---|---|
Molecular Formula | BO4P |
Molecular Weight (g/mol) | 105.78 |
MDL Number | MFCD00011318 |
InChI Key | RKVCQBPDVHFKCU-UHFFFAOYSA-N |
Synonym | boron phosphate, boron orthophosphate, boron phosphate b po4, 2,4,5-trioxa-1-phospha-3-borabicyclo 1.1.1 pentane 1-oxide, 2,4,5-trioxa-1??-phospha-3-borabicyclo 1.1.1 pentan-1-one, borophosphoric acid, boron iii phosphate, boron phosphate hydrate, boronphosphate b po4, boron phosphate 100g |
PubChem CID | 83329 |
IUPAC Name | 2,4,5-trioxa-1$l^{5}-phospha-3-borabicyclo[1.1.1]pentane 1-oxide |
SMILES | B12OP(=O)(O1)O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388457
|
Thermo Scientific Alfa Aesar
032562.22 |
100 g |
143.72 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398457
|
Thermo Scientific Alfa Aesar
032562.36 |
500 g |
581.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Boron phosphate is used as a catalyst for dehydration as well as in other organic reactions. It is employed as a source of phosphates involved in the exchange reaction to prepare metal phosphates. It finds application in refractories, ceramics and flame-retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13308-51-5 | |
105.78 | |
RKVCQBPDVHFKCU-UHFFFAOYSA-N | |
83329 | |
B12OP(=O)(O1)O2 |
BO4P | |
MFCD00011318 | |
boron phosphate, boron orthophosphate, boron phosphate b po4, 2,4,5-trioxa-1-phospha-3-borabicyclo 1.1.1 pentane 1-oxide, 2,4,5-trioxa-1??-phospha-3-borabicyclo 1.1.1 pentan-1-one, borophosphoric acid, boron iii phosphate, boron phosphate hydrate, boronphosphate b po4, boron phosphate 100g | |
2,4,5-trioxa-1$l^{5}-phospha-3-borabicyclo[1.1.1]pentane 1-oxide |
Specifications
>1400°C | |
White | |
BO4P | |
boron phosphate, boron orthophosphate, boron phosphate b po4, 2,4,5-trioxa-1-phospha-3-borabicyclo 1.1.1 pentane 1-oxide, 2,4,5-trioxa-1??-phospha-3-borabicyclo 1.1.1 pentan-1-one, borophosphoric acid, boron iii phosphate, boron phosphate hydrate, boronphosphate b po4, boron phosphate 100g | |
RKVCQBPDVHFKCU-UHFFFAOYSA-N | |
2,4,5-trioxa-1$l^{5}-phospha-3-borabicyclo[1.1.1]pentane 1-oxide | |
83329 | |
Odorless | |
Boron phosphate |
13308-51-5 | |
Powder | |
MFCD00011318 | |
Insoluble in water. | |
B12OP(=O)(O1)O2 | |
105.78 | |
105.79 | |
2.81 |
Safety and Handling
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H360FD
missing translation for 'einecsNumber' : 236-337-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only