missing translation for 'onlineSavingsMsg'
Learn More
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11376498

Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11376498
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11376498

Brand: Thermo Scientific Alfa Aesar 012134.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-42-8
Molecular Formula B
Molecular Weight (g/mol) 10.81
MDL Number MFCD00134034 MFCD00151272
InChI Key ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Synonym boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
PubChem CID 5462311
ChEBI CHEBI:27560
IUPAC Name boron
SMILES [B]

Specifications

Quantity 10 g
Percent Purity 99.5%
Odor Odorless
Particle Size 20mm (0.8 in.) & down
Form Crystalline Pieces
Assay (metals basis)
Chemical Name or Material Boron pieces

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ11J007 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number012134.30,012134.09,012134.18
Lot NumberX18H017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012134.30,012134.09,012134.18
Lot NumberE19P16A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012134.30,012134.09,012134.18
Lot NumberG11M14 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012134.30,012134.09,012134.18
Lot NumberG29M22 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012134.30,012134.09,012134.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.