missing translation for 'onlineSavingsMsg'
Learn More

Boron silicide, 98% (metals basis), Thermo Scientific Chemicals

Product Code. 11373827
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Boron silicide is used in chemical research, pharmaceuticals and as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12008-29-6
Molecular Formula B6Si
Molecular Weight (g/mol) 92.95
MDL Number MFCD00180907
InChI Key ZRBFEDMQRDRUDG-UHFFFAOYSA-N
Synonym silicon hexaboride, 1-sila-2,3,4,5,6,7-hexaborapentacyclo 3.2.0.0 1 ,?.0 2 ,?.0 3 ,? heptane, silicon hexaboride sib6 grade
PubChem CID 16212548
IUPAC Name 1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane
SMILES B12B3B4B5B(B14)[Si]235

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 92.95
Percent Purity 98%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Boron silicide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.