missing translation for 'onlineSavingsMsg'
Learn More

Boron silicide, 98% (metals basis), Thermo Scientific Chemicals

88.54 € - 355.77 €

Chemical Identifiers

CAS 12008-29-6
Molecular Formula B6Si
Molecular Weight (g/mol) 92.95
MDL Number MFCD00180907
InChI Key ZRBFEDMQRDRUDG-UHFFFAOYSA-N
Synonym silicon hexaboride, 1-sila-2,3,4,5,6,7-hexaborapentacyclo 3.2.0.0 1 ,?.0 2 ,?.0 3 ,? heptane, silicon hexaboride sib6 grade
PubChem CID 16212548
IUPAC Name 1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane
SMILES B12B3B4B5B(B14)[Si]235
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373827
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035670.09
10 g
88.54 €
10g
Please to purchase this item. Need a web account? Register with us today!
11383827
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035670.18
50 g
355.77 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Boron silicide is used in chemical research, pharmaceuticals and as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12008-29-6
92.95
ZRBFEDMQRDRUDG-UHFFFAOYSA-N
16212548
B12B3B4B5B(B14)[Si]235
B6Si
MFCD00180907
silicon hexaboride, 1-sila-2,3,4,5,6,7-hexaborapentacyclo 3.2.0.0 1 ,?.0 2 ,?.0 3 ,? heptane, silicon hexaboride sib6 grade
1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane

Specifications

12008-29-6
B6Si
silicon hexaboride, 1-sila-2,3,4,5,6,7-hexaborapentacyclo 3.2.0.0 1 ,?.0 2 ,?.0 3 ,? heptane, silicon hexaboride sib6 grade
ZRBFEDMQRDRUDG-UHFFFAOYSA-N
1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane
16212548
98%
(metals basis)
Powder
MFCD00180907
Insoluble in water.
B12B3B4B5B(B14)[Si]235
92.95
92.95
Odorless
Boron silicide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-535-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only