Learn More
Boron standard solution, for AAS, 1mg/mL B in water, Thermo Scientific Chemicals
23.52 € - 75.68 €
Specifications
Chemical Name or Material | Boron standard solution |
---|---|
Concentration | 997 to 1003 μg/mL of B in water (20°C) |
CAS | 7440-42-8 |
Packaging | Plastic bottle |
Molecular Formula | B |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10154260
|
Thermo Scientific Acros
195861000 |
100 mL | Plastic bottle |
23.52 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10504141
|
Thermo Scientific Acros
195865000 |
500 mL | Plastic bottle |
75.68 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Boron standard solution | |
7440-42-8 | |
B | |
14, 1343 | |
[B] | |
10.81 | |
CHEBI:27560 | |
AAS | |
Solution |
997 to 1003 μg/mL of B in water (20°C) | |
Plastic bottle | |
MFCD00134034 MFCD00151272 | |
ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
boron | |
5462311 | |
10.81 | |
Soluble in water |
Safety and Handling
missing translation for 'einecsNumber' : 231-151-2
missing translation for 'rtecsNumber' : ED7350000
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.