missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide dimethyl sulfide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10481854 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10481854

Brand: Thermo Scientific Acros 428321000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron tribromide dimethyl sulfide
Name Note 1M solution in methylene chloride
CAS 29957-59-3, 75-09-2
Quantity 100 mL
Molecular Formula C2H6BBr3S
MDL Number MFCD00043296
Synonym tribromoborane-methyl sulfide, boron tribromide-methyl sulfide complex, boron tribromide dimethyl sulfide complex, tribromo dimethyl-??-sulfanylidene-??-borane, dimethyl sulfide-tribromoborane, tribromo dimethylsulfonio boranuide, boron tribromide-methylsulfide complex, borontribromide-methyl sulfide complex, boron tribromide dimethyl sulfide complex solution
Solubility Information Solubility in water: reacts
InChI Key YNLFEVAOQLXINF-UHFFFAOYSA-N
SMILES CSC.BrB(Br)Br
IUPAC Name (methylsulfanyl)methane; tribromoborane
Molecular Weight (g/mol) 312.65
PubChem CID 4181510
Formula Weight 312.66
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.456
Density 1.4560g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron tribromide dimethyl sulfide, 1M solution in methylene chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.