Learn More
Boron trichloride, 1M soln. in dichloromethane, stab., Thermo Scientific Chemicals
86.73 € - 248.24 €
Chemical Identifiers
CAS | 10294-34-5 |
---|---|
Molecular Formula | BCl3 |
Molecular Weight (g/mol) | 117.16 |
MDL Number | MFCD00011313 |
InChI Key | FAQYAMRNWDIXMY-UHFFFAOYSA-N |
Synonym | boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas |
PubChem CID | 25135 |
IUPAC Name | trichloroborane |
SMILES | ClB(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408195
|
Thermo Scientific Alfa Aesar
H32751.AE |
100 mL |
86.73 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418195
|
Thermo Scientific Alfa Aesar
H32751.AP |
500 mL |
248.24 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.
Solubility
Miscible with dichloromethane, ethanol, carbon tetrachloride, diethyl ether, dimethyl formamide, aromatic solvents, saturated and halogenated hydrocarbon.
Notes
Moisture sensitive. Incompatible with water, oxidizing agents, bases and metals. Store in a cool place.
Chemical Identifiers
10294-34-5 | |
117.16 | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
25135 | |
ClB(Cl)Cl |
BCl3 | |
MFCD00011313 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
trichloroborane |
Specifications
10294-34-5 | |
MFCD00011313 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
1M soln. | |
1.326 |
BCl3 | |
UN2927 | |
Miscible with dichloromethane,ethanol,carbon tetrachloride,diethyl ether,dimethyl formamide,aromatic solvents,saturated and halogenated hydrocarbon. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
Moisture sensitive; May form a precipitate | |
Boron trichloride, stabilized |
Safety and Handling
GHS H Statement
H301-H331-H314-H351
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H331-H314-H335-H336-H350
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 233-658-4
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.