Learn More
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 198921000
64.35 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron trichloride | |
1M Solution in Hexane | |
BCl3 | |
MFCD00011313 | |
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
AcroSeal™ Glass Bottle | |
100 mL | |
Solution |
Colorless to Yellow | |
10294-34-5, 110-54-3 | |
BCl3 | |
0.9 to 1.1M | |
0.7380g/mL | |
Solubility in water: may decompose. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
0.738 | |
−17°C |
Safety and Handling
- Boron trichloride, 1M solution in hexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : Hexane, Boron trichloride
RUO – Research Use Only