missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™

Product Code. 13498598 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 13498598

Brand: Thermo Scientific Acros 445271000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 373-57-9 | BF3 | 67.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifica

Boiling Point 65.0°C
Chemical Name or Material Boron trifluoride
Melting Point -98.0°C
CAS 373-57-9, 67-56-1
Quantity 100 mL
Molecular Formula BF3
Linear Formula BF3
MDL Number MFCD00011316
Flash Point 4°C
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
Solubility Information Solubility in water: may decompose
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILES FB(F)F
IUPAC Name trifluoroborane
Molecular Weight (g/mol) 67.81
PubChem CID 11062313
Formula Weight 67.81
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.87
Density 0.8700g/mL
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • Boron trifluoride, 12% in methanol
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1
Indicazioni di pericolo
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato