missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™, Thermo Scientific Chemicals

60.70 € - 279.39 €

Specifications

Boiling Point 65.0°C
Chemical Name or Material Boron trifluoride
Melting Point -98.0°C
CAS 373-57-9, 67-56-1
Molecular Formula BF3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13498598
missing translation for 'viewDocuments'
Thermo Scientific Acros
445271000
100 mL
60.70 €
100mL
Please to purchase this item. Need a web account? Register with us today!
13408608
missing translation for 'viewDocuments'
Thermo Scientific Acros
445278000
800 mL
279.39 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

65.0°C
-98.0°C
BF3
MFCD00011316
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
WTEOIRVLGSZEPR-UHFFFAOYSA-N
trifluoroborane
11062313
AcroSeal™ Glass bottle
0.8700g/mL
Boron trifluoride
373-57-9, 67-56-1
BF3
4°C
Solubility in water: may decompose
FB(F)F
67.81
67.81
0.87
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Toxic if swallowed.
Reacts violently with water.
Toxic in contact with skin.
Causes damage to organs

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-766-4

Product Identifier
  • Boron trifluoride, 12% in methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates

RUO – Research Use Only