missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride, 12% (1.5M) in methanol, Thermo Scientific Chemicals

Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3)

30.85 € - 140.70 €

Specifications

Boiling Point 65.0°C
Chemical Name or Material Boron trifluoride
Melting Point -98.0°C
Name Note 1.5M solution in methanol
CAS 373-57-9, 67-56-1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10578122
View Documents
Thermo Scientific Acros
402761000
100 g Glass bottle
30.85 €
100g
Please to purchase this item. Need a web account? Register with us today!
10186840
View Documents
Thermo Scientific Acros
402765000
500 g Glass bottle
82.01 €
500g
Please to purchase this item. Need a web account? Register with us today!
10521944
View Documents
Thermo Scientific Acros
402760010
1 kg Glass bottle
140.70 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent, Lewis acid
Specifications

Specifications

65.0°C
-98.0°C
373-57-9, 67-56-1
Solution
BF3
4°C
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
WTEOIRVLGSZEPR-UHFFFAOYSA-N
67.81
67.81
Glass bottle
0.8700g/mL
Boron trifluoride
1.5M solution in methanol
Amber to Colorless
BF3
MFCD00011316
14, 1349
Solubility in water: may decompose
FB(F)F
11062313
10 to 15% (w/v BF3)
0.87
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Toxic if swallowed.
Reacts violently with water.
Toxic in contact with skin.
Causes damage to organs

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-766-4

missing translation for 'rtecsNumber' : ED2275000

missing translation for 'tsca' : TSCA

Product Identifier
  • Boron trifluoride, 12% in methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only