missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride diethyl etherate, 46.5% BF{3} min, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

47.54 € - 69.95 €

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
PubChem CID 8000
SMILES FB(F)F.CCOCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358958
View Documents
Thermo Scientific Alfa Aesar
041796.22
100 g
47.54 €
100g
Please to purchase this item. Need a web account? Register with us today!
11368958
View Documents
Thermo Scientific Alfa Aesar
041796.36
500 g
69.95 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

109-63-7
141.93
KZMGYPLQYOPHEL-UHFFFAOYSA-N
FB(F)F.CCOCC
C4H10BF3O
MFCD00013194
8000

Specifications

125°C to 126°C
109-63-7
C4H10BF3O
MFCD00013194
3909607
14,1350
KZMGYPLQYOPHEL-UHFFFAOYSA-N
ethoxyethane;trifluoroborane
8000
46.5% BF3 min.
ChemSeal™ bottles
Moisture sensitive
Boron trifluoride diethyl etherate
-60°C
Liquid
BF3·O(CH2CH3)2
UN2604
47°C (116°F)
Miscible with ether and alcohol.
FB(F)F.CCOCC
141.93
141.93
Acrid
1.344
1.130 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H372-H314-H318-H226-H302
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H314-H332-H372

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIFLUORIDE DIETHYL ETHERATE

missing translation for 'einecsNumber' : 203-689-8

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.