missing translation for 'onlineSavingsMsg'
Learn More

Boron triiodide, 95%, Thermo Scientific Chemicals

92.76 € - 418.08 €

Chemical Identifiers

CAS 13517-10-7
Molecular Formula BI3
Molecular Weight (g/mol) 391.52
MDL Number MFCD00036286
InChI Key YMEKEHSRPZAOGO-UHFFFAOYSA-N
Synonym boron triiodide, borane, triiodo, triiodoboron, acmc-1bo2g, boron triiodide 1g, inchi=1/bi3/c2-1 3
PubChem CID 83546
IUPAC Name triiodoborane
SMILES IB(I)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
087220.03
1 g
92.76 €
1g
Please to purchase this item. Need a web account? Register with us today!
11394949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
087220.06
5 g
418.08 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Boron triiodide is used as pharmaceutical, chemical and organic intermediate. It is also used as a catalyst and for preparation of boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13517-10-7
391.52
YMEKEHSRPZAOGO-UHFFFAOYSA-N
83546
IB(I)I
BI3
MFCD00036286
boron triiodide, borane, triiodo, triiodoboron, acmc-1bo2g, boron triiodide 1g, inchi=1/bi3/c2-1 3
triiodoborane

Specifications

210°C
13517-10-7
Crystals
BI3
boron triiodide, borane, triiodo, triiodoboron, acmc-1bo2g, boron triiodide 1g, inchi=1/bi3/c2-1 3
YMEKEHSRPZAOGO-UHFFFAOYSA-N
triiodoborane
83546
Pungent
3.35
48°C to 51°C
White
>98%
MFCD00036286
Reacts with water.
IB(I)I
391.52
391.52
Light and moisture sensitive
Boron triiodide
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II

missing translation for 'einecsNumber' : 236-857-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only