missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Borosilicate Glass 5.1 Rimless Test Tube
Manufactured from borosilicate glass.
Brand: Fisherbrand™ A510016000711
This item is not returnable.
View return policy
Description
- Borosilicate glass
- Round bottom
- Without rim
Specifications
Rimless | |
100 mm | |
Single-use | |
No |
Top style: Without rim | |
Borosilicate Glass (5.1) | |
1000 | |
16 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction