Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
251.00 € - 271.00 €
Products 3
Product Code Brand Volume (Metric) Increments Price Quantity & Availability  
Product Code Brand Volume (Metric) Increments Price Quantity & Availability  
12832104
View Documents
Pyrex
3496/04AS
25 mL 0.05 mL
261.00 €
Each
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
12842104
View Documents
Pyrex
3496/06AS
50 mL 0.1 mL
271.00 €
Each
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
12822104
View Documents
Pyrex
3496/02AS
10 mL 0.02 mL
251.00 €
Each
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
Description
Description
  • As 3396 range with intermediate stopcock
  • Intermediate stopcock facilitates easy drainage of liquid back into reservoir
  • 1.5mm bore PTFE key stopcock