Learn More
Jaytec Glass™ Borosilicate Glass Babcock Bottle
The Borosilicate Glass Babcock Bottle is used to determine the fat content of dairy samples.
Brand:  Jaytec Glass™ MP.705
 View more versions of this product
Product Code. 12338955
  • 32.65 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured in accordance with BS.755, Part 1:1937
  • Borosilicate Glass
Specifications
Specifications
Borosilicate Glass
Babcock Bottle
0 to 30%
18 g
To measure fat content in cream