missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Borosilicate Glass Base

The Borosilicate Glass Base is a replacement component part, a support base, for the Wheaton™ 90 mm Filtration Assembly. Manufactured from Wheaton™ 33 low extractable borosilicate glass conforming to USP Type I and ASTM E 438 Type I, Class A requirements.

Brand:  Wheaton™ 419455

Product Code. 11718552

  • 304.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Replacement part for 90 mm filtration assembly
  • Wheaton™ 33 low extractable borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wheaton™ 90 mm Filtration Assembly
Borosilicate Glass
1 L Funnel with Screw Collar, Glass Support Base, Filter Support, No. 8 Silicone Stopper
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.