Learn More
Fisherbrand™ Borosilicate Glass Burette Tips
Add to the end of a burette to perform volumetric titration applications. Fisherbrand™ Borosilicate Glass Burette Tips fit single bore burettes.
Brand:  Fisherbrand™ FB50055
Product Code. 12967730
  • 37.20 € / Pack of 5
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Compatible with Fisherbrand™ single bore burettes
  • Borosilicate glass
  • For use with titration applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Burette
Borosilicate Glass
Buret Clamp Tip